Learn More
Alkali Scientific Microhematocrit Capillary Tube, Glass, Blue Tip, Plain Non heparinized(Untreated), 1000/Pk, 70 µL, 75mm Length
SDP
Catalog No. 5000662375
Encompass_Preferred
Microhematocrit Capillary Tubes are manufactured from high quality precision glass to support reliable blood transfer and accurate micro hematocrit determinations in clinical and research laboratories. These plain (untreated) capillary tubes are designed for collecting anticoagulated blood directly from blood collection tubes as part of standard hematology testing workflows. The uniform internal bore promotes rapid capillary filling while preserving specimen integrity. Precision manufacturing ensures consistent dimensions and excellent sample retention during centrifugation. Tubes are suitable for micro hematocrit testing at speeds up to 14,500 RPM for approximately 5 minutes. For laboratory efficiency, the tubes are packaged with 100 capillary tubes per vial and 1,000 tubes per pack, supporting high volume hematology testing environments.
Catalog No. 50-006-62375 Supplier Alkali Scientific Supplier No. GCT70
Microhematocrit Capillary Tubes are manufactured from high quality precision glass to support reliable blood transfer and accurate micro hematocrit determinations in clinical and research laboratories. These plain (untreated) capillary tubes are designed for collecting anticoagulated blood directly from blood collection tubes as part of standard hematology testing workflows. The uniform internal bore promotes rapid capillary filling while preserving specimen integrity. Precision manufacturing ensures consistent dimensions and excellent sample retention during centrifugation. Tubes are suitable for micro hematocrit testing at speeds up to 14,500 RPM for approximately 5 minutes. For laboratory efficiency, the tubes are packaged with 100 capillary tubes per vial and 1,000 tubes per pack, supporting high volume hematology testing environments.