Learn More
Andwin Scientific MICROHEMATOCRIT CAPILLARY TUBE
SDP
Catalog No. NC9424648
Encompass_Preferred
The microhematocrit capillary tubes are made from soda lime glass. These tubes are packaged in plastic vials that have reclosable snap-lock caps to maintain its sanitation. The ends may be sealed using a flame sealing clay or patented caps.
Catalog No. NC9424648 Supplier Andwin Scientific Supplier No. B44161
The microhematocrit capillary tubes are made from soda lime glass. These tubes are packaged in plastic vials that have reclosable snap-lock caps to maintain its sanitation. The ends may be sealed using a flame sealing clay or patented caps.