Learn More
World Precision Instrument MicroImplant - Implantable Oxygen Microsensor
SDP
Catalog No. 502521406
Encompass
The MicroImplant fiber optic oxygen sensor is an implantable probe (IMP) with a tiny probe tip size 50 µm, an exposed fiber length of 5-mm and a jacket diameter of 900 µm. The IMP sensor was successfully implanted in crabs, fishes and soil.
Catalog No. 50-252-1406 Supplier World Precision Instrument Supplier No. 501658
The MicroImplant fiber optic oxygen sensor is an implantable probe (IMP) with a tiny probe tip size 50 µm, an exposed fiber length of 5-mm and a jacket diameter of 900 µm. The IMP sensor was successfully implanted in crabs, fishes and soil.