Learn More
Adaptive Science Tools Microinjection Mold I-34
Catalog No. NC1593623
Encompass
Six ramps used for micro-injection. Both sides are at a 90-degree angle firmly orienting the embryo.
Catalog No. NC1593623 Supplier Adaptive Science Tools Supplier No. I34
Six ramps used for micro-injection. Both sides are at a 90-degree angle firmly orienting the embryo.