Learn More
World Precision Instrument Microinjection Syringe Pump with SMARTouch Controller-UMP3T-1
SDP
Catalog No. 502521039
Encompass
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pumps, and/or our NANOLITER2020 positive displacement pumps. Key features of the MICRO2T controller include a splash-proof touch screen, built-in memory configured for saving settings between use, and a dual mode motor drive. The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available. 
To learn more about our warranty options, click here.
Catalog No. 50-252-1039 Supplier World Precision Instrument Supplier No. UMP3T1
The touch-screen display on WPI's MICRO2T SMARTouch™ controller offers an intuitive, user-friendly interface. Our MICRO2T allows for simultaneous control of the UMP3 syringe pumps, and/or our NANOLITER2020 positive displacement pumps. Key features of the MICRO2T controller include a splash-proof touch screen, built-in memory configured for saving settings between use, and a dual mode motor drive. The UMP3 may be mounted to either a stereotaxic frame or a micro manipulator via its built-in mounting bar. An optional foot switch is available. 
To learn more about our warranty options, click here.