Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

iLine Microsystems™ microINR Chips

Catalog No. 23241001
Change view
Click to view available options
For Use With (Equipment):
microINR Meter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-241-001 microINR Meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-241-001 Supplier iLine Microsystems™ Supplier No. CHC0025AD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Box of 25 chips for quantitative determination of prothrombin time (PT) in INR (International Normalized Ratio) units using fresh capillary blood performed by fingersticking.

  • Disposable plastic test strip
  • Fully passive (i.e., no built-in sensors, nor electrodes, nor external pumping)
  • High sensitivity human recombinant thromboplastin
  • Real control assay in parallel channel
  • Chip expiry and calibration parameters coded and integrated into the chip
  • Individually packed
  • Storage at room temperature (2°C to 25°C (36°F to 77°F))
  • 15 month shelf life

Specifications

Sample Type Fresh Capillary Blood
For Use With (Equipment) microINR Meter
Includes 25 microINR Chips
Operating System Direct Clot Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.