Learn More
iLine Microsystems microINR Expert Meter Kit
Catalog No. 22024996
Change view
Click to view available options
For Use With (Application):
Clinical and At-Home Oral Anticoagulant Therapy Monitoring
Catalog No. For Use With (Application)
22-024-996 Clinical and At-Home Oral Anticoagulant Therapy Monitoring
1 options
Catalog No. 22-024-996 Supplier iLine Microsystems Supplier No. KTQ0001AD
Add to Cart
Add to Cart
CLIA Waived PT/INR coagulometer
  • For professional use and patient self-testing
  • Comprehensive range of conductivity options (Wi-Fi, Bluetooth, Ethernet)
  • Robust traceability features and intuitive, guided user experience
  • Multilevel on-board quality control and adherence to highest safety standards
    • Ideal solution for oral anticoagulant therapy monitoring in clinical and at-home use
Specifications
Type Coagulation Analyzer Meter Kit
For Use With (Application) Clinical and At-Home Oral Anticoagulant Therapy Monitoring