Learn More
iLine Microsystems™ microINR Link Meter Kit
Catalog No. 22040059
Change view
Click to view available options
Sample Type:
Capillary Whole Blood
Catalog No. Sample Type
22-040-059 Capillary Whole Blood
1 options
Catalog No. 22-040-059 Supplier iLine Microsystems™ Supplier No. KTL0001AD
Add to Cart
Add to Cart
Point of Care device for warfarin monitoring.
  • The microINR Link Meter, included in the microINR Link Kit, is designed for use by both healthcare professionals and self-testing patients to measure INR values.
  • The meter can communicate wirelessly via Bluetooth to transfer results to compatible electronic devices such as smartphones, tablets, or computers.
  • This wireless data transfer enables convenient storage and management of results within suitable mobile applications or Data Management Software (DMS).
Specifications
Type Accuracy Reader
Sample Type Capillary Whole Blood
Dimensions (L x W x H) 119 x 65 x 35 mm