Learn More
iLine Microsystems™ microINR Starter Kit
Catalog No. 23241000
Change view
Click to view available options
Sample Type:
Fresh Capillary Blood
Catalog No. Sample Type
23-241-000 Fresh Capillary Blood
1 options
This item is not returnable. View return policy
Catalog No. 23-241-000 Supplier iLine Microsystems™ Supplier No. SKA0001AD
Add to Cart
Add to Cart
This item is not returnable. View return policy
The microINR starter kit includes one meter and 6 boxes of 25 chips.
  • Small blood sample required (3 μL)
  • Direct application to test chip means that no pipettes are needed
  • Automatic Strip Identification (no need to match lot numbers)
  • Auto Calibration means parameters are encoded within each individual chip
  • On-board and Internal Controls in both chip and monitor
  • No need of external liquid controls
Specifications
Type Coagulometer
Sample Type Fresh Capillary Blood
Includes 1 microINR Meter, 6 Boxes of microINR Chips
Operating System Direct Clot Detection