Learn More
Gfg Instrumentation MICROIV CHLOR 0-10.0PPM 0.1PPM
SDP
Catalog No. NC3734748
Encompass_Preferred
Micro IV / Chlorine (Cl2) / 0 - 10.0 ppm / 0.1 ppm resolution - Does not include calibration adapter P/N 7770-201C
Catalog No. NC3734748 Supplier Gfg Instrumentation Supplier No. 1418153
Micro IV / Chlorine (Cl2) / 0 - 10.0 ppm / 0.1 ppm resolution - Does not include calibration adapter P/N 7770-201C