Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System
Catalog No. 14816244 Shop All Hamilton Company Products
Change view
Click to view available options
Type:
Dual Syringe Continuous Dispenser
Dual Syringe Diluter
Dual Syringe Dispenser
Single Syringe Dispenser
Controller Type:
Advanced Controller
Basic Controller
DTHP
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Controller Type
14-816-244 Dual Syringe Diluter Advanced Controller
14-816-243 Single Syringe Dispenser Advanced Controller
14-816-246 Dual Syringe Continuous Dispenser Advanced Controller
14-816-239 Single Syringe Dispenser Basic Controller
14-816-242 Dual Syringe Continuous Dispenser Basic Controller
14-816-241 Dual Syringe Dispenser Basic Controller
14-816-245 Dual Syringe Dispenser Advanced Controller
14-818-142 Dual Syringe Diluter DTHP
14-816-240 Dual Syringe Diluter Basic Controller
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-816-244 Supplier Hamilton™ Supplier No. ML625DIL
Add to Cart
Add to Cart
Request a Quote
Dilute/Dispense with better than 99% accuracy, independent of a liquid's viscosity, vapor pressure, and temperature. Microlab™ 600 Diluter/Dispenser System is a highly precise syringe pump with a graphical user interface designed to quickly and easily dilute and dispense volumes from 10μL to 50mL.
  • Available as a single or dual syringe system
  • High torque, precision stepper motors deliver positional accuracy
  • Supports syringes from 10μL to 50mL
  • Ethernet port for communication with controller or corporate network
  • Printer Kit available
Syringe Pump Features
  • High torque valve motors
  • Precision syringe drives with 48,000 step resolution over 60 minutes
  • Lighted power and prime buttons
  • Independent left and right trigger ports
  • Fanless heat vent
  • 24V power input
  • CAN Daisy Chain input/out
  • RS-232 console port
  • Power over Ethernet (PoE)
  • TTL Input/Output
Dual Syringe Diluter
  • Creates up to a 1:50,000 dilution in single step
  • Reduces preparation time and wasted buffer
  • Washes tubing between each sample
Dispenser/Diluter Printer Kit
  • Prints on standard, non-thermal 8 1/2 x 11 and A4 paper
  • Interfaces with the free compliance software package that is available for Advanced Microlab™ devices
  • Satisfies regulatory CFR21 Part 11 and FDA GMP/GLP requirements by conveniently printing all log files directly from the device's controller
Dispenser:
Sample spiking, Titration, Animal dosing (oral gavage), Reagent addition to chemical reactors, Flow chemistry, Manufacturing e.g. Mercury dispensing for light bulbs, Ink cartridge filling, Glue and epoxy dispensing, Nasal therapeutics (viscous), Contact lenses
Diluter:
Forensics blood alcohol analysis and urinalysis, minig assay labs metals detection, environmental analysis water and soils testing, oil analysis preventative maintenance, alcohol testing beer and wine manufacturing
Order Info
Microlab 600 Printer Kit sold separately
Specifications
Type Dual Syringe Diluter
Controller Type Advanced Controller