Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System
Shop All Hamilton Company ProductsDescription
- Available as a single or dual syringe system
- High torque, precision stepper motors deliver positional accuracy
- Supports syringes from 10μL to 50mL
- Ethernet port for communication with controller or corporate network
- Printer Kit available
Syringe Pump Features
- High torque valve motors
- Precision syringe drives with 48,000 step resolution over 60 minutes
- Lighted power and prime buttons
- Independent left and right trigger ports
- Fanless heat vent
- 24V power input
- CAN Daisy Chain input/out
- RS-232 console port
- Power over Ethernet (PoE)
- TTL Input/Output
Dual Syringe Diluter
- Creates up to a 1:50,000 dilution in single step
- Reduces preparation time and wasted buffer
- Washes tubing between each sample
Dispenser/Diluter Printer Kit
- Prints on standard, non-thermal 8 1/2 x 11 and A4 paper
- Interfaces with the free compliance software package that is available for Advanced Microlab™ devices
- Satisfies regulatory CFR21 Part 11 and FDA GMP/GLP requirements by conveniently printing all log files directly from the device's controller
Dispenser:
Sample spiking, Titration, Animal dosing (oral gavage), Reagent addition to chemical reactors, Flow chemistry, Manufacturing e.g. Mercury dispensing for light bulbs, Ink cartridge filling, Glue and epoxy dispensing, Nasal therapeutics (viscous), Contact lenses
Diluter:
Forensics blood alcohol analysis and urinalysis, minig assay labs metals detection, environmental analysis water and soils testing, oil analysis preventative maintenance, alcohol testing beer and wine manufacturing
Order Info
Microlab 600 Printer Kit sold separately
Specifications
Specifications
Type | Dual Syringe Diluter |
Controller Type | Advanced Controller |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.