Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 11mm AlumiTin Magnetic Crimp Top Autosampler Vial Seals
SDP

Catalog No. 15111485 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Silver
Yellow
Septum:
FEP/Butyl
FEP/Natural Rubber
PTFE/Silicone
PTFE/Silicone/PTFE
Quantity:
100
2000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum Quantity
15-111-485 Silver FEP/Natural Rubber 100
15-111-505 Yellow FEP/Butyl 100
15-111-519 Silver PTFE/Silicone 100
15-111-520 Silver PTFE/Silicone 2000
15-111-525 Silver PTFE/Silicone/PTFE 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15-111-485 Supplier DWK Life Sciences Supplier No. 110020AT
Add to Cart
Add to Cart

MicroLiter's 11mm AlumiTin crimp-top autosampler vial seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) FEP/Natural Rubber
Color Silver
Material Aluminum
Pre-slit No
Septum FEP/Natural Rubber
Quantity 100
Closure Size 11 mm
For Use With (Equipment) 11 mm Autosampler Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.