Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter™ 11 mm Crimp Top Autosampler Vial Seals
SDP

Catalog No. 15111507 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-111-507 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-111-507 Supplier DWK LIFE SCIENCES™ Supplier No. 110022AB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter™ 11 mm Crimp Top Autosampler Vial Seals are designed for exceptional sealing performance with volatile samples. Available in multiple colors and septa types to suit a range of autosampler applications.

  • 11 mm crimp top seals designed for use with MicroLiterTM autosampler vials
  • Available in multiple colors for easy sample identification
  • Multiple septa types available to suit a range of analytical applications
  • Provides exceptional sealing performance for volatile samples
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Diameter (Metric) Outer 11 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum ETFE, Silicone
Product Line MicroLiter
Quantity 2000/Pk.
Closure Size 11 mm
For Use With (Equipment) Autosampler Vials
Autoclavable Not Autoclavable
Product Type Crimp Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.