Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 16 mm Screw Thread Headspace Vial Caps
SDP

Catalog No. 15111614 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
Blue PTFE, White Silicone
FKM
None
PTFE, Butyl
PTFE, Silicone
White PTFE, Blue Silicone
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15-111-614 FKM
15-111-604 None
15-111-607 PTFE, Butyl
15-111-609 White PTFE, Blue Silicone
15-111-610 PTFE, Silicone
15-111-612 Blue PTFE, White Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15-111-614 Supplier DWK LIFE SCIENCES™ Supplier No. 160080M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 16 mm Screw Thread Headspace Vial Caps are designed for use with 16 mm headspace vials and are available in multiple septa types, including PTFE/Silicone, PTFE/Butyl, Silicone/Aluminum, and FKM, to suit a range of chromatography applications.

  • 16 mm screw thread design for secure, reliable closure on headspace vials
  • Available in multiple septa types including PTFE/Silicone, PTFE/Butyl, Silicone/Aluminum, FKM, and No Septum options
  • Silver aluminum cap construction
  • Suitable for headspace sampling and chromatography applications including GC and GC-MS
  • Available in pack sizes of 100 and 1000
  • Manufactured by MicroLiter, a DWK Life Sciences brand
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Diameter (Metric) Outer 16 mm
Material Steel
Pre-slit No
Sterility Non-sterile
Septum FKM
No. per Pack 100
Product Line MicroLiter
Quantity 100/Pk.
Closure Size 16 mm
For Use With (Equipment) 16 mm Screw Thread Headspace Vials - MicroLiter
Autoclavable Not Autoclavable
Product Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.