Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 20 mm Aluminum Crimp Top Headspace Vial Seals, No Liner
SDP

Catalog No. 15111627 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
Silver
No. per Pack:
100
1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
15-111-627 Silver 100
15-111-629 Blue 100
15-111-633 Green 100
15-111-628 Silver 1000
15-111-641 Red 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15-111-627 Supplier DWK LIFE SCIENCES™ Supplier No. 200000AS
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 20 mm Aluminum Crimp Top Headspace Vial Seals, No Liner are designed for use with headspace sampling vials and are available in multiple colors and pack sizes. These aluminum crimp top seals provide a reliable, liner-free closure for chromatography applications.

  • 20 mm aluminum crimp top seals designed for headspace sampling vials
  • No liner construction for direct septum contact applications
  • Compatible with all standard headspace autosampler systems (except LEAP/CTC)
  • Available in multiple colors for easy sample identification and organization
  • Available in standard (100/pack) and bulk (1000/pack) quantities
  • Suitable for use with a range of septa types when paired with appropriate septum components
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum None
No. per Pack 100
Product Line MicroLiter
Quantity 100/Pk.
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
Autoclavable Not Autoclavable
Product Type Crimp Seal
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.