Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm Aluminum Crimp Top Headspace Vial Seals, No Liner
SDP

Catalog No. p-7237556 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
Silver
No. per Pack:
100
1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
15-111-629 Blue 100
15-111-633 Green 100
15-111-641 Red 100
15-111-627 Silver 100
15-111-628 Silver 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15-111-629 Supplier DWK Life Sciences Supplier No. 200000B
Only null left
Add to Cart
Add to Cart

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Aluminum
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.