Learn More
DWK LIFE SCIENCES™ MicroLiter 20 mm AlumiTin Crimp Top Headspace Vial Seals
Shop All DWK Life Sciences Products

Description
- 20 mm AlumiTin crimp top seals designed for headspace vial applications
- Available in multiple septa types including Butyl, PTFE/Butyl, Natural PTFE/Blue Silicone, and Tan PTFE/White Silicone
- Color-coded aluminum caps allow easy visual identification and sample tracking
- Compatible with a wide range of headspace autosamplers
- Supplied in packs of 100 for convenient laboratory use
- MicroLiter brand quality from DWK Life Sciences
Specifications
Specifications
| Color | Silver |
| Diameter (Metric) Outer | 20 mm |
| Material | Aluminum |
| Pre-slit | No |
| Sterility | Non-sterile |
| Septum | PTFE/Butyl |
| No. per Pack | 100 |
| Product Line | MicroLiter |
| Quantity | 100/Pk. |
| Closure Size | 20 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.