Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm AlumiTin Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111717 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Red
Silver
Septum:
Natural PTFE, Blue Silicone
PTFE, Butyl
Tan PTFE, White Silicone
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15-111-717 Silver Natural PTFE, Blue Silicone
15-111-688 Silver PTFE, Butyl
15-111-709 Blue Natural PTFE, Blue Silicone
15-111-719 Red Natural PTFE, Blue Silicone
15-111-736 Silver Tan PTFE, White Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15-111-717 Supplier DWK Life Sciences Supplier No. 200050NT
Add to Cart
Add to Cart

MicroLiter 20 mm crimp top seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Material Aluminum
Pre-slit No
Septum Natural PTFE, Blue Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.