Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20mm AlumiTin Large Opening Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111741 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
Aluminum/Silicone
Butyl stopper
Molded PTFE/Butyl
Natural PTFE/Blue Silicone
PTFE/Butyl
Tan PTFE/White Silicone
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15-111-741 Aluminum/Silicone
15-111-679 PTFE/Butyl
15-111-692 Molded PTFE/Butyl
15-111-704 Natural PTFE/Blue Silicone
15-111-725 Tan PTFE/White Silicone
15-111-674 Butyl stopper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15-111-741 Supplier DWK Life Sciences Supplier No. 200060AT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 20mm large opening crimp top seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Aluminum/Silicone
Color Green
Material Aluminum
Pre-slit No
Septum Aluminum/Silicone
Quantity 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.