Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm AlumiTin Small Opening Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111705 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
Natural PTFE, Blue Silicone
Tan PTFE, White Silicone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15-111-705 Natural PTFE, Blue Silicone
15-111-726 Tan PTFE, White Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-111-705 Supplier DWK Life Sciences Supplier No. 200050ATS
Add to Cart
Add to Cart

MicroLiter 20 mm small opening crimp top seals are available in multiple colors and septa types.

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Aluminum
Pre-slit No
Septum Natural PTFE, Blue Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.