Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 20 mm AlumiTin Small Opening Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111726 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
Natural PTFE, Blue Silicone
Tan PTFE, White Silicone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15-111-726 Tan PTFE, White Silicone
15-111-705 Natural PTFE, Blue Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-111-726 Supplier DWK LIFE SCIENCES™ Supplier No. 200051ATS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 20 mm AlumiTin Small Opening Crimp Top Headspace Vial Seals are compatible with LEAP and CTC systems. Available in multiple septa types including Butyl, PTFE/Butyl, and Silicone combinations. Supplied in packs of 100.

  • 20 mm AlumiTin small opening crimp top design for headspace vials
  • Compatible with LEAP and CTC autosampler systems
  • Available in multiple septa types: Butyl, PTFE/Butyl, Molded PTFE/Butyl, Natural PTFE/Blue Silicone, Tan PTFE/White Silicone, and Aluminum/Silicone
  • Supplied in packs of 100
  • AlumiTin construction provides consistent crimp performance
  • Small opening format suitable for headspace sampling applications
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum Tan PTFE, White Silicone
No. per Pack 100
Product Line MicroLiter
Quantity 100/Pk.
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Autoclavable Not Autoclavable
Product Type Crimp Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.