Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 20 mm Bi-Metal Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111631 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
Septum:
Aluminum/Silicone
Natural PTFE, Blue Silicone
None
PTFE/Butyl
PTFE/Silicone
Tan PTFE, White Silicone
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Septum
15-111-631 Green None
15-111-681 Blue PTFE/Butyl
15-111-706 Blue Natural PTFE, Blue Silicone
15-111-707 Green Natural PTFE, Blue Silicone
15-111-708 Red Natural PTFE, Blue Silicone
15-111-727 Blue Tan PTFE, White Silicone
15-111-728 Green PTFE/Silicone
15-111-745 Red Aluminum/Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 15-111-631 Supplier DWK LIFE SCIENCES™ Supplier No. 200000BMG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 20 mm Bi-Metal Crimp Top Headspace Vial Seals are designed for use with 20mm crimp top headspace vials. Available in multiple colors and septa types including Butyl, PTFE/Butyl, PTFE/Silicone, and Aluminum/Silicone for flexible compatibility.

  • 20mm bi-metal crimp top seals designed for headspace vial applications
  • Available in multiple cap colors including blue, green, and red for easy sample identification
  • Multiple septa options available: Butyl, PTFE/Butyl, Molded PTFE/Butyl, PTFE/Silicone, and Aluminum Foil/Silicone
  • PTFE-faced septa provide a highly chemical resistant barrier
  • Compatible with aluminum open top crimp seals
  • Designed for 20mm vial mouth ODs
  • Supplied in packs of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Diameter (Metric) Outer 20 mm
Material Bi-metal
Pre-slit No
Sterility Non-sterile
Septum None
No. per Pack 100
Product Line MicroLiter
Quantity 100/Pk.
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
Autoclavable Not Autoclavable
Product Type Crimp Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.