Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers
SDP

Catalog No. 15111739 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Blue, Natural
Gray
White
White, Tan
Material:
Aluminum, Silicone
Butyl
Molded PTFE, Butyl
PTFE, Butyl
PTFE, Silicone
Product Type:
3-leg Stopper
Septa
Stopper
No. per Pack:
100
1000
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Product Type No. per Pack
15-111-739 White Aluminum, Silicone Septa 100
15-111-657 Black Butyl Septa 100
15-111-677 Gray PTFE, Butyl Septa 100
15-111-690 Gray Molded PTFE, Butyl Septa 100
15-111-702 Blue, Natural PTFE, Silicone Septa 100
15-111-721 White, Tan PTFE, Silicone Septa 100
15-111-722 White, Tan PTFE, Silicone Septa 1000
15-111-670 Gray Butyl Stopper 100
15-111-671 Gray Butyl Stopper 1000
15-111-672 Gray Butyl 3-leg Stopper 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 15-111-739 Supplier DWK Life Sciences Supplier No. 200060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers are designed for use with 20 mm crimp seal headspace vials. Butyl stoppers suit applications where PTFE is not required; PTFE/butyl and molded PTFE/butyl septa deliver low background peaks and a tight seal.

  • Designed for use with WHEATON MicroLiter 20 mm crimp seal headspace vials
  • Butyl-only and butyl stopper options available for applications where PTFE is not required
  • PTFE/butyl septa provide an economical choice with low background peaks
  • Molded PTFE/butyl septa offer the tightest seal of all PTFE items and low background peaks
  • PTFE/silicone septa are excellent for volatile compound analysis
  • Aluminum foil/silicone septa are suitable for volatile applications
  • Multiple material combinations available to match instrument and application requirements
  • Available in standard packs of 100 and bulk packs of 1000
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Aluminum, Silicone
Pre-slit No
Sterility Non-sterile
No. per Pack 100
Quantity 100/Cs.
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Seals - MicroLiter
Autoclavable Not Autoclavable
Product Type Septa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.