Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20 mm Crimp Top Headspace Vial Septas and Stoppers
SDP

Catalog No. 15111643 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-111-643 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-111-643 Supplier DWK Life Sciences Supplier No. 200020

missing translation for 'validMainframeMsg'

Butyl stoppers can be used when PTFE is not required. Butyl/PTFE septa are an economical choice with low background peaks.

Molded PTFE/butyl septa offer the tightest seal of all PTFE items, as well as low background peaks. PTFE/silicone septa are excellent for volatiles. Aluminum/silicone septa are also excellent for volatiles, but not suitable for repeat injections.
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Butyl
Pre-slit No
No. per Pack 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Seals - MicroLiter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.