Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 20mm Magnetic Large Opening Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111714 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Gold
Silver
Material (Liner):
Butyl
Natural PTFE/Blue Silicone
PTFE/Butyl
Tan PTFE/White silicone
Quantity:
100
1000
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material (Liner) Quantity
15-111-714 Gold Natural PTFE/Blue Silicone 100
15-111-685 Silver PTFE/Butyl 100
15-111-712 Silver Natural PTFE/Blue Silicone 100
15-111-715 Gold Natural PTFE/Blue Silicone 1000
15-111-731 Silver Tan PTFE/White silicone 100
15-111-732 Silver Tan PTFE/White silicone 1000
15-111-733 Gold Tan PTFE/White silicone 100
15-111-734 Gold Tan PTFE/White silicone 1000
15-111-653 Gold Butyl 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 15-111-714 Supplier DWK Life Sciences Supplier No. 200050MLS
Add to Cart
Add to Cart

MicroLiter 20mm crimp top seals are available in multiple colors and septa types.

Magnetic large opening crimp seal
TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Natural PTFE/Blue Silicone
Color Gold
Material Steel
Pre-slit No
Septum PTFE/Silicone
Quantity 100
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Vials - MicroLiter
Product Type Crimp Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.