Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 20 mm Pressure Release Crimp Top Headspace Vial Seals
SDP

Catalog No. 15111640 Shop All DWK Life Sciences Products
Change view
Click to view available options
Septum:
Aluminum/Silicone
None
PTFE/Butyl
Tan PTFE/White Silicone
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
15-111-640 None
15-111-689 PTFE/Butyl
15-111-737 Tan PTFE/White Silicone
15-111-748 Aluminum/Silicone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-111-640 Supplier DWK LIFE SCIENCES™ Supplier No. 200000P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 20 mm Pressure Release Crimp Top Headspace Vial Seals are designed for use with headspace sampling systems and are available in multiple septa types. These aluminum crimp top seals feature a pressure release mechanism and are compatible with most autosampler platforms.

  • 20 mm aluminum crimp top seals designed for headspace vial applications
  • Pressure release design activates at 5.0 Bar for safe headspace sampling
  • Available in multiple septa types including Butyl, PTFE/Butyl, PTFE/Molded Butyl, PTFE/Silicone, and Aluminum/Silicone
  • Compatible with most headspace autosampler systems (excluding LEAP/CTC)
  • Supplied in quantities of 100 per pack
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum None
No. per Pack 100
Product Line MicroLiter
Quantity 100/Cs.
Closure Size 20 mm
For Use With (Equipment) 20 mm Headspace Septa - MicroLiter
Product Type Pressure Release Crimp Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.