Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 9 mm ScrewTin Autosampler Vial Caps with FEP/Natural Rubber Septa
SDP

Catalog No. 15111030 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-111-030 Red
15-111-018 Blue
15-111-021 Green
15-111-033 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-111-030 Supplier DWK LIFE SCIENCES™ Supplier No. 090020RT
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 9 mm ScrewTin Autosampler Vial Caps with FEP/Natural Rubber Septa provide a reliable seal for 9 mm screw-thread autosampler vials. Available in multiple cap colors to support color-coded sample organization. Supplied in packs of 100.

  • ScrewTin cap design for use with 9 mm screw-thread autosampler vials
  • FEP/Natural Rubber/FEP septa construction for reliable sample containment
  • Available in multiple cap colors: Black, Blue, Green, Natural, Red, and Yellow for easy color-coded sample identification
  • Supplied in packs of 100
  • DWK Life Sciences MicroLiter brand quality
TRUSTED_SUSTAINABILITY

Specifications

Color Red
Diameter (Metric) Outer 9 mm
Material Tinplate Steel
Pre-slit No
Sterility Non-sterile
Septum FEP/Natural Rubber/FEP
No. per Pack 100
Product Line MicroLiter
Quantity 100/Pk.
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Autoclavable Not Autoclavable
Product Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.