Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9 mm ScrewTin Autosampler Vial Caps with PTFE/Silicone Septa
SDP

Catalog No. p-7237563 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Yellow
Pre-slit:
No
Yes
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Pre-slit
15-111-074 Black No
15-111-068 Blue No
15-111-088 Blue Yes
15-111-071 Green No
15-111-077 Natural No
15-111-095 Natural Yes
15-111-083 Yellow No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 15-111-074 Supplier DWK Life Sciences Supplier No. 090030KT
Only null left
Add to Cart
Add to Cart

Available in multiple colors and package sizes.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polypropylene
Pre-slit No
Septum PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.