Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ MicroLiter 9mm Screwthread Autosampler Vial Caps with Open Tops
SDP

Catalog No. 15111013 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Blue
Green
Natural
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
15-111-013 Yellow
15-111-007 Blue
15-111-008 Green
15-111-009 Black
15-111-010 Natural
15-111-012 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15-111-013 Supplier DWK LIFE SCIENCES™ Supplier No. 090000Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter 9mm Screwthread Autosampler Vial Caps with Open Tops feature an open-top design for use with autosampler systems. Available in multiple colors for easy sample identification. Compatible septa sold separately. Sold in packs of 100.

  • 9mm screw-thread design for secure closure on compatible autosampler vials
  • Open-top configuration allows septum insertion for needle access
  • Available in multiple colors for easy sample identification and organization
  • Compatible with MicroLiter 9mm screwthread autosampler vial septa, sold separately
  • Sold in packs of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Diameter (Metric) Outer 9 mm
Material Polypropylene
Pre-slit No
Sterility Non-sterile
Septum None
Quantity 100/Pk.
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Autoclavable Not Autoclavable
Product Type Open Top Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.