Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter Glass Limited Volume Inserts for 9 mm Autosampler Vials, Untreated
SDP

Catalog No. 15111003 Shop All DWK Life Sciences Products
Change view
Click to view available options
No. per Pack:
100
1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
15-111-003 100
15-111-004 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-111-003 Supplier DWK Life Sciences Supplier No. 090000101
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

MicroLiter Glass Limited Volume Inserts for 9 mm Autosampler Vials, Untreated are made from Type 1 borosilicate glass and meet strict tolerances. Flat bottom, pulled point, and plastic spring-loaded styles stay vertical in the vial for consistent low-volume sampling.

  • Manufactured from Type 1 borosilicate glass for chemical compatibility and sample integrity
  • Sized to fit the inside diameter of 9 mm autosampler vial necks, keeping inserts vertical during sampling
  • Available in flat bottom, pulled point (conical), and plastic spring-loaded designs to suit varied process workflows
  • Offer the maximum opening of any insert on the market for reliable low-volume sample access
  • Untreated surface suitable for standard analytical applications
  • Compatible with most autosampler platforms using 9 mm vials
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Borosilicate Glass (Type 1)
Sterility Non-sterile
Volume (Metric) 350 μL
No. per Pack 100
Volume (Metric) Insert 350 μL
Base Style Conical with Pulled Point
Product Line MicroLiter
Quantity 100/Cs.
Marking Spot No
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Autoclavable Autoclavable
Product Type Conical Insert
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.