Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One MICROLON™ 200 12-well Elisa Strips

Catalog No. 07000771 Shop All Greiner Bio-One Products
Change view
Click to view available options
MICROLON™ 200 12-Well Elisa Strips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
07-000-771 MICROLON™ 200 12-Well Elisa Strips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-771 Supplier Greiner Bio-One Supplier No. 701001

missing translation for 'validMainframeMsg'

For use in research. Greiner Bio-One MICROLON™ 200 12-Well Elisa Strips offer flexibility in diagnostics.

  • Polystyrene
  • Medium binding
  • 1 x 12 well
  • Flat bottom strips for Plate Frame 701185
  • Clear
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Product Type Elisa Strip
Material Polystyrene
No. of Wells 12
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
For Use With (Application) ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.