Learn More
Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates
Catalog No. 07000772 Shop All Greiner Bio-One Products
Change view
Click to view available options
Binding Property:
Medium Binding
Catalog No. Binding Property
07-000-772 Medium Binding
1 options
Catalog No. 07-000-772 Supplier Greiner Bio-One Supplier No. 701070
For use in flexibility in diagnostics. Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates are suitable for transmission measurements.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • For optical measurements
  • Measuring light beam is not deflected by well profile
  • Specifically treated to provide incresed protein binding
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Strip Plate
Array 8 x 12
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Quantity 100/Cs.