Learn More
Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates
Catalog No. 07000773 Shop All Greiner Bio-One Products
Change view
Click to view available options
Binding Property:
High Binding
Medium Binding
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Binding Property
07-000-773 High Binding
07-000-772 Medium Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-773 Supplier Greiner Bio-One Supplier No. 701071
For use in flexibility in diagnostics. Greiner Bio-One MICROLON™ 200 96-well Polystyrene Strip Plates are suitable for transmission measurements.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • For optical measurements
  • Measuring light beam is not deflected by well profile
  • Specifically treated to provide incresed protein binding
TRUSTED_SUSTAINABILITY
Specifications
Binding Property High Binding
Format Strip Plate
Array 8 x 12
Binding Type High Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Quantity 100/Cs.