Learn More
Greiner Bio-One MICROLON™ 200 Flat-Bottom 96-well Microplate
GREENER_CHOICE
Catalog No. 07000099 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-099 Clear
1 options
Catalog No. 07-000-099 Supplier Greiner Bio-One Supplier No. 655080
Add to Cart
Add to Cart
Free of detectable DNase, RNase, human DNA and pyrogens
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Non-sterile
Surface Treatment Microlon 200
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Well Color Clear
Well Design Chimney
Dimensions (L x W x H) 127.76 x 85.48 x 14.40 mm