Learn More
Greiner Bio-One MICROLON™ 600 High Binding 96-well ELISA Assay Microplates
GREENER_CHOICE
Catalog No. 07000627 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-000-627 Clear
1 options
Catalog No. 07-000-627 Supplier Greiner Bio-One Supplier No. 655085
Add to Cart
Edge
Add to Cart
For consistent performance in binding assays. Greiner Bio-One MICROLON™ 600 High Binding 96-Well ELISA Assay Microplates are free of detectable DNAse, RNase and human DNA.
  • Made of optically clear polystyrene
  • Supplied without lid
  • Chimney-well design helps reduce cross-contamination
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Microlon 600
Well Shape Round, C Bottom
Quantity 40/Cs.