Learn More
Greiner Bio-One MICROLON™ 600 High-Binding Single-Break Strip Plate
Catalog No. 07001022 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
Catalog No. Color
07-001-022 Clear
1 options
Catalog No. 07-001-022 Supplier Greiner Bio-One Supplier No. 70507125
Add to Cart
Add to Cart
Allows individual wells to be broken off separately, allowing number of tests to be adjusted to number of samples.
  • High binding clear strips
  • Made from polystyrene
  • C-Bottom, flat with curved edge
  • Non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Breakapart Single break
Format Standard
Description strip plate
Array 8 x 12
Binding Type High Binding
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Well Shape Curved Edges, Flat Bottom (C-well)
Well Color Clear