Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
GREENER_CHOICE

Catalog No. 07000777 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Blue, Clear Bottom
Clear
Clear, Blue
Clear, Green
Clear, Red
Clear, Yellow
Green, Clear Bottom
Red, Clear Bottom
Binding Property:
High Binding
Medium Binding
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Binding Property
07-000-777 Green, Clear Bottom Medium Binding
07-000-775 Red, Clear Bottom Medium Binding
07-000-776 Blue, Clear Bottom Medium Binding
07-000-779 Clear Medium Binding
07-000-780 Clear High Binding
07-000-781 Clear, Red High Binding
07-000-782 Clear, Blue High Binding
07-000-783 Clear, Green High Binding
07-000-784 Clear, Yellow High Binding
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 07-000-777 Supplier Greiner Bio-One Supplier No. 705065
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in colorimetric immunoassay. Greiner Bio-One MICROLON™ 96-Well ELISA Single-Break Strip Plates can be broken off separately and number of tests performed can be adjusted to number of samples.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Individual wells can be pipetted without leaving residue
  • Flat bottom enables precise optical measurements
  • Suitable for transmission measurements
  • Specifica
TRUSTED_SUSTAINABILITY

Specifications

Breakapart Single break
Binding Property Medium Binding
Format Strip Plate
Array 8 x 12
Binding Type Medium Binding
Color Green, Clear Bottom
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon
Well Shape Curved Edges, Flat Bottom (C-well)
Dnase Free Yes
For Use With (Application) Colorimetric immunoassay
Quantity 100/Cs.
RNase Free Yes
Dimensions (L x W x H) 127.76 x 85.48 x 14.5 mm (5.02 x 3.36 x 0.5 in.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.