Learn More
Key Scientific Products MICROLOOPS PLASTIC 5UL LOOPS
SDP
Catalog No. NC2120591
Encompass_Preferred
MICROLOOPS PLASTIC 5 uL LOOPS INDIVIDUALLY PACKAGED: Sterile plastic inoculating loops. Great for general microbiology applications.Features:Precision moulded.Standard shaft.Individually packed with 1 loop in each sterile peel pouch.Benefits:No gougingThe single packs are ideal for special projects or academic use
Catalog No. NC2120591 Supplier Key Scientific Products Supplier No. MW705/1
MICROLOOPS PLASTIC 5 uL LOOPS INDIVIDUALLY PACKAGED: Sterile plastic inoculating loops. Great for general microbiology applications.Features:Precision moulded.Standard shaft.Individually packed with 1 loop in each sterile peel pouch.Benefits:No gougingThe single packs are ideal for special projects or academic use