Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Catalog No. 524490 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
HM 525
HM 550 and 560
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
52-449-0 HM 525
14-070-88 HM 550 and 560
28-21 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 52-449-0 Supplier Epredia™ Supplier No. 524490
Add to Cart
Add to Cart
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
Specifications
Product Type Heat Extractor
For Use With (Equipment) HM 525