Learn More
MSC Proto Torque Wrench, 1/2 in. Drive, 70-350 Nm, 1 ea
Catalog No. 501919946
Encompass
This micrometer torque wrench features a 1/2 in. drive and a ratcheting head, measuring torque in Newton Meters. It is manufactured for standard industrial torque applications, heavy equipment, and general maintenance.
  • Calibrated to ±3% clockwise and ±6% counter clockwise from 20% to 100% of full scale
  • Calibration is accomplished by manual loading of the torque wrench
  • Meets ASME B107.14M-1994 standards
  • Constructed from high-grade steel with heat-treated critical parts for durability
  • Features a torque range of 70–350 Nm with 1 Nm increments
  • Overall length of 27-1/8 in.
Catalog No. 50-191-9946 Supplier MSC Supplier No. 38362182
This micrometer torque wrench features a 1/2 in. drive and a ratcheting head, measuring torque in Newton Meters. It is manufactured for standard industrial torque applications, heavy equipment, and general maintenance.
  • Calibrated to ±3% clockwise and ±6% counter clockwise from 20% to 100% of full scale
  • Calibration is accomplished by manual loading of the torque wrench
  • Meets ASME B107.14M-1994 standards
  • Constructed from high-grade steel with heat-treated critical parts for durability
  • Features a torque range of 70–350 Nm with 1 Nm increments
  • Overall length of 27-1/8 in.