Learn More
MSC Proto Micrometer Torque Wrench, 3/4 in, 160-800 Nm, 1 ea
Catalog No. 501920095
Encompass
This Micrometer Torque Wrench is designed for precise torque applications, featuring a ratcheting head for efficient operation.
  • Has a 3/4 in drive
  • Measures torque in newton meters
  • Minimum torque of 160 Nm
  • Maximum torque of 800 Nm
  • Graduation of 4 Nm
  • Overall length of 41-9/16 in
  • Features a ratcheting head
Catalog No. 50-192-0095 Supplier MSC Supplier No. 38362216
This Micrometer Torque Wrench is designed for precise torque applications, featuring a ratcheting head for efficient operation.
  • Has a 3/4 in drive
  • Measures torque in newton meters
  • Minimum torque of 160 Nm
  • Maximum torque of 800 Nm
  • Graduation of 4 Nm
  • Overall length of 41-9/16 in
  • Features a ratcheting head