Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ MICROMAN™ E Capillary Pistons PROMO

Catalog No. F148414GPM Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
Sterility:
Non-sterile
Sterile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
F148414GPM 10 to 100 μL Non-sterile
F148415GPM 10 to 100 μL Sterile
F148413GPM 1 to 10 μL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. F148414GPM Supplier Gilson™ Supplier No. F148414PM
Get Up to 50% Off Gilson Liquid Handling Solutions
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.

  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.

Specifications

Volume (Metric) 10 to 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model CP100
Length (English) 4.8 in.
Length (Metric) 41.8 mm
Format Box
Tip Style Positive Displacement
Packaging Box
Quantity 192 Pk.
Product Type Capillary Piston
Disposable Single-use
For Use With (Equipment) Gilson Microman
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.