Learn More
World Precision Instrument Micromanifold, 250 µm ID Tip
SDP
Catalog No. 502521334
Encompass
The most unique feature of the MPS-2 is its perfusion micromanifold. Using the latest microfluidic techniques, the injection molded micromanifold provides the least flow resistance and dead volume of any product on the market. The flow channel inner diameter is approximately 1.0 mm, except for the last 5 mm before the junction point. This design allows a fast flow rate without using a pressured system. Small channels and a unique design at the merging point further reduce the chance of cross contamination. Dead volume is less than 100 nL at junction.
Catalog No. 50-252-1334 Supplier World Precision Instrument Supplier No. 502125
The most unique feature of the MPS-2 is its perfusion micromanifold. Using the latest microfluidic techniques, the injection molded micromanifold provides the least flow resistance and dead volume of any product on the market. The flow channel inner diameter is approximately 1.0 mm, except for the last 5 mm before the junction point. This design allows a fast flow rate without using a pressured system. Small channels and a unique design at the merging point further reduce the chance of cross contamination. Dead volume is less than 100 nL at junction.