Learn More
Sutter Instruments Micromanipulators
SDP
Catalog No. NC9819363
Encompass_Preferred
Right-handed micromanipulator with vertical lock with 1/2 inch rod clamps. The MM-33 is the right choice for tight environments which require maximum versatility within a small space. Stainless steel cross-loaded roller bearings are designed to offer low friction and smooth linear motion with a minimum of side play. The pre-loaded rollers are set at 90 degrees to the hardened steel guides insuring constant contact and accuracy. This precise rack and pinion drive gives stable, drift-free movement with minimal backlash. The controls are placed one above the other which makes for less hand movement and easier positioning. The scales for all axes are calibrated in 0.10mm increments and the fine micrometer adjustment for the X-axis thrust is calibrated at 0.01mm with estimates to 5μm.
Catalog No. NC9819363 Supplier Sutter Instruments Supplier No. FGMM33R
Right-handed micromanipulator with vertical lock with 1/2 inch rod clamps. The MM-33 is the right choice for tight environments which require maximum versatility within a small space. Stainless steel cross-loaded roller bearings are designed to offer low friction and smooth linear motion with a minimum of side play. The pre-loaded rollers are set at 90 degrees to the hardened steel guides insuring constant contact and accuracy. This precise rack and pinion drive gives stable, drift-free movement with minimal backlash. The controls are placed one above the other which makes for less hand movement and easier positioning. The scales for all axes are calibrated in 0.10mm increments and the fine micrometer adjustment for the X-axis thrust is calibrated at 0.01mm with estimates to 5μm.