Learn More
World Precision Instrument Kite Manual Micromanipulator-KITE-M3-R
SDP
Catalog No. NC0095313
Encompass
The KITE is an economical manual micromanipulator that is perfect for student use. It is available in right or left hand versions and is also sold bundled with an optional M3 Tilting Base. Our 5 lb. weight (WPI #5464) is frequently sold with this unit to provide stability.
Catalog No. NC0095313 Supplier World Precision Instrument Supplier No. KITEM3R
The KITE is an economical manual micromanipulator that is perfect for student use. It is available in right or left hand versions and is also sold bundled with an optional M3 Tilting Base. Our 5 lb. weight (WPI #5464) is frequently sold with this unit to provide stability.