Learn More
Molecular Devices Molecular Devices Low-Volume Slide Refill Kit, 64-Well, Pack of 3
Catalog No. 5000014505
Encompass_Preferred
Replenishment kit of low-volume bottom slides for slide-based microplate readers. The kit includes three low-volume bottom slides, each with 64 wells, intended for low-volume absorbance and related assays.
  • Contains three 64-well low-volume bottom slides.
  • Intended for low-volume absorbance and slide-based assays.
  • Supplies replacement slides to restore slide reader capacity.
Catalog No. 50-000-14505 Supplier Molecular Devices Supplier No. 02006264
Replenishment kit of low-volume bottom slides for slide-based microplate readers. The kit includes three low-volume bottom slides, each with 64 wells, intended for low-volume absorbance and related assays.
  • Contains three 64-well low-volume bottom slides.
  • Intended for low-volume absorbance and slide-based assays.
  • Supplies replacement slides to restore slide reader capacity.