Learn More
Molecular Devices Molecular Devices SpectraDrop Cover Slides, 4 μL, 1.0 mm Path
Catalog No. 5000014363
Encompass_Preferred
Molecular Devices SpectraDrop Micro-Volume cover slide refills provide replacement cover slides for low-volume absorbance and fluorescence measurements. Each pack contains three 4 μL cover slides with a 1.0 mm path length, designed to restore and extend micro-volume workflows while minimizing sample consumption.
  • Pack contains three 4 μL cover slides.
  • Path length 1.0 mm.
  • Enables measurements with sample volumes as low as 2 μL.
  • Designed for micro-volume absorbance and fluorescence measurements.
  • Helps reduce sample consumption and increase throughput.
Catalog No. 50-000-14363 Supplier Molecular Devices Supplier No. 02006266
Molecular Devices SpectraDrop Micro-Volume cover slide refills provide replacement cover slides for low-volume absorbance and fluorescence measurements. Each pack contains three 4 μL cover slides with a 1.0 mm path length, designed to restore and extend micro-volume workflows while minimizing sample consumption.
  • Pack contains three 4 μL cover slides.
  • Path length 1.0 mm.
  • Enables measurements with sample volumes as low as 2 μL.
  • Designed for micro-volume absorbance and fluorescence measurements.
  • Helps reduce sample consumption and increase throughput.