Learn More
Molecular Devices Molecular Devices SpectraDrop Micro-Volume Microplate Kit, High-Throughput, 2/4 μL
Catalog No. 5000014538
Encompass_Preferred
The SpectraDrop MV HTS Kit is a high-throughput micro-volume microplate kit for low-volume quantitation of DNA, RNA, and protein using 2 μL and 4 μL sample volumes. The kit provides an adapter, multiple low-volume slides in 24- and 64-well formats, top cover slides with defined path lengths, and a protective carrying case for storage and transport.
  • High-throughput micro-volume solution for low-volume measurements (2 μL and 4 μL)
  • Includes adapter, five 24-well bottom slides, five 64-well bottom slides, and multiple top cover slides
  • Teflon-coated bottom slides for precise droplet placement and easy cleaning
  • Top slides with side spacers to control pathlength and reduce evaporation
  • Compatible with absorbance and fluorescence detection for DNA, RNA, and protein quantitation
  • Supplied in a protective carrying case for storage and transport
Catalog No. 50-000-14538 Supplier Molecular Devices Supplier No. 02006267
Add to Cart
Add to Cart
Request a Quote
The SpectraDrop MV HTS Kit is a high-throughput micro-volume microplate kit for low-volume quantitation of DNA, RNA, and protein using 2 μL and 4 μL sample volumes. The kit provides an adapter, multiple low-volume slides in 24- and 64-well formats, top cover slides with defined path lengths, and a protective carrying case for storage and transport.
  • High-throughput micro-volume solution for low-volume measurements (2 μL and 4 μL)
  • Includes adapter, five 24-well bottom slides, five 64-well bottom slides, and multiple top cover slides
  • Teflon-coated bottom slides for precise droplet placement and easy cleaning
  • Top slides with side spacers to control pathlength and reduce evaporation
  • Compatible with absorbance and fluorescence detection for DNA, RNA, and protein quantitation
  • Supplied in a protective carrying case for storage and transport