Learn More
Molecular Devices Molecular Devices SpectraDrop Refill Kit, 2 μL Cover, 3 Slides
Catalog No. 5000014367
Encompass_Preferred
Replacement micro-volume cover slides supplied as a refill kit for SpectraDrop micro-volume sample holders. The kit provides precision top slides for low-volume absorbance and fluorescence measurements, enabling reproducible quantitation from 2 μL samples.
  • Contains three 2 μL cover slides (0.5 mm path length).
  • Designed for micro-volume absorbance and fluorescence measurements.
  • Enables accurate DNA, RNA, and protein quantitation from 2 μL samples.
  • Disposable slides reduce cross-contamination risk.
  • Facilitates rapid sample preparation and higher throughput.
Catalog No. 50-000-14367 Supplier Molecular Devices Supplier No. 02006265
Replacement micro-volume cover slides supplied as a refill kit for SpectraDrop micro-volume sample holders. The kit provides precision top slides for low-volume absorbance and fluorescence measurements, enabling reproducible quantitation from 2 μL samples.
  • Contains three 2 μL cover slides (0.5 mm path length).
  • Designed for micro-volume absorbance and fluorescence measurements.
  • Enables accurate DNA, RNA, and protein quantitation from 2 μL samples.
  • Disposable slides reduce cross-contamination risk.
  • Facilitates rapid sample preparation and higher throughput.